Sterilization Indicator Tape
Sterilization Indicator Tape
Actual product may vary
Fisherbrand
Sterilization Indicator Tape
Indicates sterilization with simple color change of white to brown. Kartell™ Sterilization Indicator Tape is used with autoclaving or dry heat sterilization methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilization Method
FKS10245041Poupinel Method (Dry Heat)
FKS10073454Autoclave
Catalog number FKS10245041
Price (SGD)
181.00
EA
Add to cart
Sterilization Method:
Poupinel Method (Dry Heat)
Price (SGD)
181.00
EA
Add to cart
Indicates sterilization with simple color change of white to brown. Kartell™ Sterilization Indicator Tape is used with autoclaving or dry heat sterilization methods.
  • Adhesive tape indicates sterilization
  • Tapes available for autoclaving and Poupinet methods of sterilization:
    • Autoclave tape: 20’ at +121°C or 10’ at +125°C or 5’ at +135°C
    • Poupinel method: dry heat
  • Color changes from white to brown
  • Lead free if correctly stocked
  • Autoclave tape can be used until 3 years from production date
  • Poupinel tape can be used until 24 months from production date
  • Conformity with ISO 11140/2005
Specifications
Certifications/ComplianceISO 11140/2005
For Use With (Application)To indicate sterilization
Length (Metric)50 m
MaterialPaper
Sterilization IndicatorYes
Sterilization MethodPoupinel Method (Dry Heat)
Width (Metric) Exterior19 mm
Unit SizeEA