CellBIND™ Surface Microplates
CellBIND™ Surface Microplates
Actual product may vary
Fisherbrand
CellBIND™ Surface Microplates
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
Have Questions?
Catalog NumberCulture AreaCapacity (Metric)Volume (Metric) WorkingWell Count
FKS102348329.5 cm22.9 mL (Well)1.9 to 2.9 mL6
Catalog number FKS10234832
Price (SGD)
1,378.00
PK
Add to cart
Culture Area:
9.5 cm2
Capacity (Metric):
2.9 mL (Well)
Volume (Metric) Working:
1.9 to 2.9 mL
Well Count:
6
Price (SGD)
1,378.00
PK
Add to cart
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Specifications
Closure ColorClear
Closure IncludedYes
Closure MaterialPolystyrene
Bottom ShapeFlat
Capacity (Metric)2.9 mL (Well)
ColorClear
Culture Area9.5 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
Surface TreatmentCellBIND
TypeMulti-well Cell Culture Plate
Volume (Metric) Working1.9 to 2.9 mL
Well Count6
IncludesLid
No. per Case50/Cs.
No. per Pack5/Pk.
Unit SizePK