ONE™ Oil Baths
ONE™ Oil Baths
Actual product may vary
Fisher Catalogue
ONE™ Oil Baths
Protect laboratory and samples, even at high temperatures. Memmert™ ONE™ Oil Baths offers multiple temperature protection through corrosion-resistant stainless steel and precise electronics.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1023326322 L
Catalog number FKS10233263
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
22 L
A range of test tube racks, baskets and other accessories is available, please contact Customer Service.
  • Fuzzy supported PID microprocessor controller
  • Digital display
  • 2 x PT100 sensors
  • Calibration facility on controller
  • Audio and visual alarm
  • Over-temperature protection
Specifications
Controller TypeFuzzy-supported PID-microprocessor controller
Capacity (Metric)22 L
Certifications/ComplianceCE, VDE, GS
DisplayDigital LED
For Use With (Application)For mining, food and beverage, temperature control
MaterialStainless Steel
ModelONE 22
Temperature (Metric) Maximum200°C
Temperature Resolution±0.1°C
Length (Metric)578 mm
Length (Metric) Interior350 mm
Width (Metric)436 mm
Width (Metric) Interior290 mm
Height (Metric)296 mm
Height (Metric) Interior220 mm
Depth (Metric) Interior350 mm
Shipping Weight (Metric)20 kg
Weight (Metric)16 kg
Voltage230 V
Wattage2 w