Manual Crimpers, Decrimpers and Decrimping Pliers
Manual Crimpers, Decrimpers and Decrimping Pliers
Actual product may vary
Manual Crimpers, Decrimpers and Decrimping Pliers
Manual Crimpers, Decrimpers and Decrimping Pliers
Fisherbrand
Manual Crimpers, Decrimpers and Decrimping Pliers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeType
FKS1022996432 mmManual Crimper
FIS03-375-711 mmManual Crimper
FIS03-375-17A13 mmManual Crimper
FIS03-375-75C13 mmManual Crimper (Flip-off)
FKS1023158513 mmManual Crimper (Flip-off)
FIS03-375-24A20 mmManual Crimper
FIS03-375-75D20 mmManual Crimper (Flip-off)
FIS03-375-75B32 mmManual Crimper
FKS1518878420 mm
FIS03-375-8A11 mmManual Decrimper
FIS03-375-24AC20 mmManual Decrimper
FIS03-377-12532 mmManual Decrimper
FIS03-375-811 mmManual Decapping Plier
Catalog number FKS10229964
Price (SGD)
1,609.00
EA
Add to cart
Size:
32 mm
Type:
Manual Crimper
Price (SGD)
1,609.00
EA
Add to cart
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Manual Crimpers
Lightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
  • Factory set to properly crimp standard-thickness seals
  • Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decrimpers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
  • Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
  • Minimizes damage to vial rim with chromatography or serum vials
  • Removes standard aluminum seals and many steel seals
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionManual Crimper, Standard
TypeManual Crimper
For Use With (Equipment)Standard aluminum caps
Size32 mm
For Use With (Application)Industrial Chromatography
Product LineNational
Unit SizeEA