Matrix™ Reagent Reservoirs
Actual product may vary
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Matrix™ Reagent Reservoirs
Fisherbrand
Matrix™ Reagent Reservoirs
Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
Have Questions?
Catalog NumberCapacity (Metric)MaterialPackagingType
FKS1022574325 mLPolystyrene10 Bags of 10Reagent Reservoir with Divider
Catalog number FKS10225743
Price (SGD)
597.00
Pack
Capacity (Metric):
25 mL
Material:
Polystyrene
Packaging:
10 Bags of 10
Type:
Reagent Reservoir with Divider
Price (SGD)
597.00
Pack
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
  • Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
Specifications
ColorWhite
Capacity (Metric)25 mL
MaterialPolystyrene
SterilitySterile
TypeReagent Reservoir with Divider
Packaging10 Bags of 10
Unit SizePack