1x1 image pixel for data collection
MiniMix 100 P CC. 100 mL Lab Blender
MiniMix 100 P CC. 100 mL Lab Blender
Actual product may vary
Fisherbrand
MiniMix 100 P CC. 100 mL Lab Blender
The MiniMix 100 P CC is made for blending small samples. Great tool for labs.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1022318180 mL
Catalog number FKS10223181
Price (SGD)
19,340.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
80 mL
Price (SGD)
19,340.00
EA
Add to cart
100 mL lab homogenizer for sample preparation:
  • Plain door
  • Multi-function digital screen
  • Useful volume: 5 - 80 mL
  • Variable speed (3 - 6 - 9 - 12 strokes/s)
  • Blending timer (1s-1hor∞)
  • Adjustable blending power (patented)
  • Click & Clean™ removable paddles (patented)
  • Side-by-side paddle stop
  • Q-Tight™ closing: tight closing for no more leaks from the top of the bag
  • Fully opening door (270°) Delivered with: 1 power cable / 1 user’s manual / 1 quick user guide / 1 free pack of blender bags
Specifications
Product TypeLab Blender
Voltage110/220 V
Electrical Requirements110/220 V, 50/60 Hz
Hertz50/60 Hz
Wattage110 W
Capacity (Metric)80 mL
Volume Range (Metric)50 to 80 mL
Depth (English)14.57 in.
Depth (Metric)37 cm
Dimensions (L x W x H)20 x 36.9 x 20.2 cm
Height (English)7.87 in.
Height (Metric)20 cm
Length (English)14.53in
Width (English)7.87 in.
Width (Metric)21 cm
Base MaterialStainless Steel
Container MaterialStainless Steel
Certifications/ComplianceISO 7218, ISO 6887 and FDA BAM (Bacteriological Analytical Manual)
DescriptionThe MiniMix 100 P CC is made for the blending of small samples. The patented adjustable blending power enables optimal bacterial extraction, and can be adapted to the nature of the sample. The MiniMix 100 P CC is equipped with patented Click & Clean click-on paddles that allow full access to the blending chamber for cleaning.
For Use With (Application)BSE tests, biopsy analyses, veterinary analyses, HPLC, GCMS, etc.
For Use With (Equipment)sample bags
IncludesDelivered with: 1 power cable, user manual, quick user guide, 1 free pack of blender bags
ModelMiniMix 100 P CC
No. of Strokes3, 6, 9, 12 strokes/sec.
Packaging Quantity1
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection