Polysine Adhesion Slides
Product Image
Fisherbrand
Polysine Adhesion Slides
Epredia™TM Polysine adhesion slides are electrostatically and biochemically adhesive. The preparation is first adhered by electrostatic attraction and then fixed in place by biochemical binding. Polysine adhesion slides are best suited to paraffin-embedded sue slices from human sources fixed in formalin, alcohol or Bouin solution. For tissue slices 2 to 5 micrometers thick.
Have Questions?
Catalog number FKS10219280
Price (SGD)
77.00
PK
Add to cart
Price (SGD)
77.00
PK
Add to cart
  • No time wasted on slide-coating proceduress
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
Specifications
Length (Metric)75 mm
Width (Metric)25 mm