Sterile Spatulas, V-scoop
Sterile Spatulas, V-scoop
Actual product may vary
Fisherbrand
Sterile Spatulas, V-scoop
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™ Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
Have Questions?
Catalog NumberLength (Metric)
FKS1021605124.9 cm
Catalog number FKS10216051
Price (SGD)
1,190.00
PK
Add to cart
Length (Metric):
24.9 cm
Price (SGD)
1,190.00
PK
Add to cart
  • Corning spatulas are designed to save time and to provide contamination-free samples.
  • Eliminates the recycling and resterilizing necessary with reusable spatulas.
  • Available in five different configurations.
  • Microspatulas available in two configurations.
  • Individually packaged.
  • Antistatic.
  • Sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Specifications
SterilitySterile
Certifications/ComplianceUSP Class VI, non-pyrogenic, RNase and DNase-free Certified
DisposableSingle-use
Product TypeSpatula
For Use With (Application)Molecular biology applications, lyophilized samples and contamination-free sampling
Length (English)9.8 in.
IncludesV-scoop at one end and a larger round spoon at the other end
MaterialPolystyrene
Length (Metric)24.9 cm
Unit SizePK