McMaster Counting Chamber
McMaster Counting Chamber
Actual product may vary
Fisherbrand
McMaster Counting Chamber
Use Marienfeld Superior™ Mc Master counting chambers for counting worm eggs
Have Questions?
Catalog NumberDimensions (L x W)
FKS1019602375 x 32mm
Catalog number FKS10196023
Price (SGD)
909.00
EA
Add to cart
Dimensions (L x W):
75 x 32mm
Price (SGD)
909.00
EA
Add to cart
  • For counting worm eggs
  • Complete set with cover glass
Specifications
Product TypeCounting Chamber
DescriptionWith 2 counting grids
ModelMcMaster
Quantity1
Dimensions (L x W)75 x 32mm
Depth (Metric)1.5mm
For Use With (Application)Suitable for solid media
IncludesWith cover glass
Unit SizeEA