Oxoid™ Rifampcin Antimicrobial Susceptibility discs
Oxoid™ Rifampcin Antimicrobial Susceptibility discs
Actual product may vary
Oxoid™ Rifampcin Antimicrobial Susceptibility discs
Oxoid™ Rifampcin Antimicrobial Susceptibility discs
Fisherbrand
Oxoid™ Rifampcin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Have Questions?
Catalog NumberConcentration
FKS1018819230 μg
Catalog number FKS10188192
Price (SGD)
-
Concentration:
30 μg
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Rifampcin Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Antimicrobial AgentsRifampcin
Certifications/ComplianceComplies with SFM guidelines
Concentration30 μg
DescriptionOxoid Rifampcin Antimicrobial Susceptibility Discs
Antibiotic CodeRD
Product TypeAntimicrobial Susceptibility Discs
Test TypeAntimicrobial Susceptibility
No. per Pack5 Cartridges/Pk.
Packaging Type50 Discs/Cartridge
Quantity50 Discs/Cartridge
Unit SizePK