18/10 Stainless Steel Micro Spoon Spatulas
Actual product may vary
Fisherbrand
18/10 Stainless Steel Micro Spoon Spatulas
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas serve a wide variety of functions in the lab.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Length (Metric) BladeWidth (Metric) Blade
FKS10186310150 mm9 mm (spoon) / 40 mm (blade)5 mm (spoon) / 5 mm (blade)
FKS11825061150 mm9 mm (spoon) / 40 mm (blade)5 mm (spoon) / 5 mm (blade)
Catalog number FKS10186310
Price (SGD)
-
Length (Metric):
150 mm
Length (Metric) Blade:
9 mm (spoon) / 40 mm (blade)
Width (Metric) Blade:
5 mm (spoon) / 5 mm (blade)
  • 18/10 Stainless steel
Specifications
ShapeSpoon End, Blade End
Product TypeMicro Spoon Spatula
Length (Metric) Blade9 mm (spoon) / 40 mm (blade)
Width (Metric)5 mm
Width (Metric) Blade5 mm (spoon) / 5 mm (blade)
Dimensions (L x W)150 x 5 mm
MaterialStainless Steel (18/10)
Length (Metric)150 mm
Unit SizeEach