Bacti-Cinerator IV Infrared Sterilizer
Bacti-Cinerator IV Infrared Sterilizer
Actual product may vary
Fisherbrand
Bacti-Cinerator IV Infrared Sterilizer
Sterilize items by incinerating organic materials using infrared heat. Cole-Parmer™ Bacti-Cinerator IV Infrared Sterilizer requires only 5-7 seconds for optimum sterilization at 1500°F.
Have Questions?
Catalog NumberSterilization Time
FKS101807305 to 7 sec.
Catalog number FKS10180730
Price (SGD)
5,232.00
EA
Add to cart
Sterilization Time:
5 to 7 sec.
Price (SGD)
5,232.00
EA
Add to cart
Sterilize items by incinerating organic materials using infrared heat. Cole-Parmer™ Bacti-Cinerator IV Infrared Sterilizer requires only 5-7 seconds for optimum sterilization at 1500°F.
  • Uses infrared heat to incinerate organic material deep within the ceramic funnel
  • Sterilizes platinum loops and needles safely and conveniently
  • Prevents infectious spatter and cross contamination
  • Sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500°F
  • Unit can be used in anaerobic chambers
  • This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications
Specifications
Product TypeBacterial Incinerator/Infrared Sterilizer
Temperature (English)1500°F
Frequency50/60 Hz
Sterilization Time5 to 7 sec.
Sterilization MethodInfrared heat
Voltage220 VAC
Unit SizeEA