Rack for Conical Bottom Centrifuge Tubes
Rack for Conical Bottom Centrifuge Tubes
Actual product may vary
Rack for Conical Bottom Centrifuge Tubes
Rack for Conical Bottom Centrifuge Tubes
Fisherbrand
Rack for Conical Bottom Centrifuge Tubes
For conical bottom tubes
Have Questions?
Catalog NumberPositionsHolds
FKS1016421225Tubes (50 mL), Tubes (29.1 mm)
Catalog number FKS10164212
Price (SGD)
142.00
Pack
Add to cart
Positions:
25
Holds:
Tubes (50 mL), Tubes (29.1 mm)
Price (SGD)
142.00
Pack
Add to cart
These polystyrene foam racks are designed to hold conical bottom plastic centrifuge tubes.
Specifications
AutoclavableNot Autoclavable
HoldsTubes (50 mL), Tubes (29.1 mm)
MaterialPolystyrene
Positions25
SterilityNon-sterile
For Use With (Equipment)Conical bottom centrifuge tubes
No. per Case20/Cs.
No. per Pack20/Pk.
SizeHolds (25) 50mL conical-bottom plastic centrifuge tubes
Product TypeCentrifuge Tube Rack
Unit SizePack