Sedgewick Rafter Counting Chamber
Sedgewick Rafter Counting Chamber
Actual product may vary
Fisherbrand
Sedgewick Rafter Counting Chamber
Counting Chambers Sedgewick Rafter are intended for counting of particles and microorganisms in water or other transparent liquids.
Have Questions?
Catalog NumberDepth (Metric)
FKS101598221 mm
Catalog number FKS10159822
Price (SGD)
2,438.00
EA
Add to cart
Depth (Metric):
1 mm
Price (SGD)
2,438.00
EA
Add to cart
  • The cell of 50 x 20 x 1mm is ruled in a 1mm grid subdividing 1 mL in 1000 μl
  • Supplied with cover glass of approx. 60 x 30 x 1mm
  • Available as precision type made of glass and a single type made of plastic
Specifications
Product TypeCounting Chamber
DescriptionPrecision type with chromium deposited for phase contrast use with ground and chamfered edges
MaterialGlass
ModelSedgewick Rafter
Depth (Metric)1 mm
For Use With (Application)Intended for Counting of Particles and Microorganisms in Water or Other Transparent Liquids
IncludesCell of 50 x 20 x 1mm ruled in a 1mm grid
Unit SizeEA