epT.I.P.S.™ Standard Pipette Tips
Actual product may vary
Fisherbrand
epT.I.P.S.™ Standard Pipette Tips
Use these universal pipette tips with a wide variety of pipette models. Eppendorf™ epT.I.P.S.™ Standard Pipettor Tips feature optimal wetting properties, high transparency and special, certified purity grades.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)Color CodeColor
FKS10157291100 to 5000 μL120 mmVioletColorless
FKS103336821000 to 10,000 μL165 mmTurquoiseColorless
FKS101991711000 to 10,000 μL243 mmTurquoiseColorless
Catalog number FKS10157291
Price (SGD)
345.00
Pack
Volume (Metric):
100 to 5000 μL
Length (Metric):
120 mm
Color Code:
Violet
Color:
Colorless
Price (SGD)
345.00
Pack
Use these universal pipette tips with a wide variety of pipette models. Eppendorf™ epT.I.P.S.™ Standard Pipettor Tips feature optimal wetting properties, high transparency and special, certified purity grades.
  • Minimal attachment and ejection forces with the highest level of tightness
  • The ergonomically optimized cone geometry provides the optimal pipette fit on the Eppendorf pipette
  • Fine graduations make visual checks of the pipetting volume even easier
  • Fine retracted or extended tips simplify work with small volumes
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
Specifications
Volume (Metric)100 to 5000 μL
SterilitySterile
ColorColorless
AutoclavableAutoclavable
Length (English)4.7 in.
Length (Metric)120 mm
FormatBulk
Tip StyleUniversal
Packaging500 Tips (5 Bags x 100 Tips)
Quantity500 Pk.
Product TypePipette Tips
Color CodeViolet
For Use With (Equipment)Eppendorf, Universal
GraduatedYes
Product LineepT.I.P.S.
GraduationsFine
Unit SizePack