Transferpette™ S Single Channel Adjustable Pipet
Transferpette™ S Single Channel Adjustable Pipet
Actual product may vary
Fisherbrand
Transferpette™ S Single Channel Adjustable Pipet
Exhibits a high degree of accuracy, precision, maximum versatility, and reliable performance with Easy Calibration™ technology.Transferpette™ S Single Channel Adjustable Pipet offers simple one-handed operation with an adjustable brake to protect against accidental volume changes.
Have Questions?
Catalog NumberVolume (Metric)Compatible TipsIncrementsIncludes
FKS10155862500 to 5000 μL0.5 to 5mL tip5μLPerformance certificate, operating manual, shelf mount, and three year warranty.
Catalog number FKS10155862
Price (SGD)
-
Volume (Metric):
500 to 5000 μL
Compatible Tips:
0.5 to 5mL tip
Increments:
5μL
Includes:
Performance certificate, operating manual, shelf mount, and three year warranty.
The Transferpette™ S Single Channel Adjustable Pipet offers robust yet lightweight construction, simple, one-handed operation, a high degree of accuracy and precision with Easy Calibration™ technology for long lasting reliability. The single channel model is available in fixed volume and digital models in volume ranges from 0.1μL to 10mL.
  • Centralized pipetting button with optimized stroke length
  • Adjustable brake to fix volume range
  • Visible four-digit volume display
  • Compatible with universal tips (0.1-1μL requires nano-cap tips)
  • Repeatedly autoclavable at 121°C/250°F(20min.)
Specifications
Accuracy±0.6%
Certifications/ComplianceCE, IVD
Compatible Tips0.5 to 5mL tip
AutoclavableAutoclavable
For Use With0.5 to 5 mL Tip
ModelD-5000
Number of Channels1
Product LineTransferpette™ S
TypeSingel Channel Pipette
Volume (Metric)500 to 5000 μL
DisplayDigital
Finger StyleCurved
For Use With (Application)Demanding laboratory applications
For Use With (Equipment)0.5 to 5mL tip
IncludesPerformance certificate, operating manual, shelf mount, and three year warranty.
Increments5μL
Warranty3 Years
Unit SizePK