Costar™ Ultra-Low Attachment Microplates
Costar™ Ultra-Low Attachment Microplates
Actual product may vary
Fisherbrand
Costar™ Ultra-Low Attachment Microplates
Featuring a covalently bound hydrogel layer that effectively inhibits cellular attachmment. Corning™ Costar™ Ultra-Low Attachment Microplates greatly reduce binding of attachment and serum proteins, minimizing cell attachment and spreading.
Have Questions?
Catalog NumberDiameter (Metric)
FKS101544313.48 cm
Catalog number FKS10154431
Price (SGD)
2,725.00
PK
Add to cart
Diameter (Metric):
3.48 cm
Price (SGD)
2,725.00
PK
Add to cart
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Specifications
Array2 x 3
Closure ColorClear
Closure IncludedYes
Closure MaterialPolystyrene
Bottom ShapeFlat
Capacity (Metric)2.9 mL (Well)
ColorClear
Culture Area9.5 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
TypeMulti-well Cell Culture Plate
Well Count6
For Use With (Application)Maintaining cells in a suspended, unattached state
IncludesLid
No. per Case24/Cs.
No. per Pack1/Pk.
PackagingIndividually Wrapped
Quantity24/Cs.
Diameter (Metric)3.48 cm
Unit SizePK