Orion™ 2-Cell Conductivity Electrodes
Orion™ 2-Cell Conductivity Electrodes
Actual product may vary
Fisherbrand
Orion™ 2-Cell Conductivity Electrodes
Achieve accurate measurements from ultrapure water, sea water, and more with these versatile 2-cell conductivity electrodes, available in a variety of materials and measurement ranges.
Have Questions?
Catalog NumberCell ConstantMeasurement RangesConnector Types
FKS101530121 cm^-11 μS/cm to 200 mS/cm8-pin Waterproof DIN
Catalog number FKS10153012
Price (SGD)
2,995.00
EA
Add to cart
Cell Constant:
1 cm^-1
Measurement Ranges:
1 μS/cm to 200 mS/cm
Connector Types:
8-pin Waterproof DIN
Price (SGD)
2,995.00
EA
Add to cart
The Thermo Scientific™ Orion™ Conductivity electrode 013016MD complies with SM 2510B; D1125-95, -99 (A) for Drinking water measurements and USP<645> Measuring the Conductivity of Pure Water.
Features
These probes use two cell electrodes to measure low-, standard-, and high-conductivity samples based on the cell constant. They are a versatile option for precise measurements in the lab and field.
013016MD is ideal in low ionic strength solutions, such as deionized water and ultrapure water when paired with a flow-cell assembly . 018020MD is good for chemically reactive conductivity samples, including concentrated acids and bases. 011050MD and 011510MD epoxy/graphite cells are durable, chemical resistant, and measure a wide range of conductivities in the lab or field.
2-cell conductivity electrodes are compatible with Thermo Scientific™ Orion Star™ , Lab Star™, Pro Star™, Versa Star Pro™ bench and portable conductivity meters. Paired together the instruments can be used to generate accurate, repeatable readings for conductivity, resistivity, TDS and salinity.
Solutions required
Standards
011008         100 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100910     147 µS/cm conductivity standard, 10 pouches
011007         1413 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100710     1413 µS/cm conductivity/TDS standard, 10 pouches
011006         12.9 mS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100610     12.9 mS/cm conductivity/TDS standard, 10 pouches
011005         111 mS/cm conductivity standard, 5 x 60 mL bottles
01100510     111 mS/cm conductivity standard, 10 pouches
Conditioning solution
011001         Conditioning solution for 011050MD conductivity cells
Accessories
013017         Replacement flow cell
Items included with
011050MD
011007         1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
011001         Conditioning solution, 60 mL 1 bottle
011510MD
011006         12.9 mS/cm conductivity/TDS standard, 60 mL 1 bottle
011007         1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
013016MD
011008         100 µS/cm conductivity/TDS standard, 60 mL 1 bottle
013017         Replacement flow cell
Warranty period
12 months from date of purchase
Specifications
For Use With (Equipment)555A, 550A, 162A, 136A, 135A, 130A Meters
Cell Constant1 cm^-1
Type2-Electrode Conductivity Cells
Connector Types8-pin Waterproof DIN
DescriptionOrion 2 Cell Chemically Resistant Conductivity Probe
MaterialGlass, Platinum
Measurement Ranges1 μS/cm to 200 mS/cm
Unit SizeEA