Heavy-Duty Test-Tube Racks
Heavy-Duty Test-Tube Racks
Actual product may vary
Fisherbrand
Heavy-Duty Test-Tube Racks
Heavy-duty 3-tier test tube racks have hand grips in the end plates for easy transport.
Have Questions?
Catalog NumberPositionsHoldsArray
FKS1014992072Tubes (10 to 13 mm)6 x 12
Catalog number FKS10149920
Price (SGD)
259.00
EA
Add to cart
Positions:
72
Holds:
Tubes (10 to 13 mm)
Array:
6 x 12
Price (SGD)
259.00
EA
Add to cart
  • Stackable, compact and steam autoclavable at 121°C (250°F) for durability in the lab
  • End plates have hand grips for easy transport
  • Base tier has rounded wells with drain holes, and the top tier has lettered and numbered rows
  • Available in different sizes
  • 8.3 x 4.3 in. (21.3 x 11 cm)
Specifications
Product TypeTest Tube Rack
Array6 x 12
AutoclavableAutoclavable
ColorTranslucent
HoldsTubes (10 to 13 mm)
MaterialPolypropylene
Positions72
ShapeRectangular
Shelves3 Tiers
StackableNo
SterilityNon-sterile
For Use With (Equipment)Heavy-duty test tube
Dimensions (L x W x H)8.37 x 4.31 x 4.37 in. (21.3 x 11 x 11.1 cm)
Length (Metric)21.3 cm
Length (English)8.37 in.
Width (Metric)11 cm
Width (English)4.31 in.
Height (Metric)11.1 cm
Height (English)4.37 in.
Diameter (Metric) Opening13 mm
Unit SizeEA