Check Order Status
Stepped Microtip
Actual product may vary
Fisherbrand
Stepped Microtip
Robust and reliable microtips and probes that intensify and radiate ultrasonic energy into a sample.
Have Questions?
Catalog Number
Diameter (Metric)
FKS10131492
6 mm
Catalog number
FKS10131492
Price (SGD)
1,607.00
EA
Add to cart
Diameter (Metric):
6 mm
Price (SGD)
1,607.00
EA
Add to cart
Microtip probe, 2 mm, 0.1 mL to 5 mL
Microtips and probes intensify and radiate the ultrasonic energy into the sample
Smaller diameter tips produce greater intensity of cavitation, but the energy released is restricted to a narrower, more concentrated field
Conversely, larger diameter tips produce lower intensity, but the energy is released over a greater area
The larger the tip diameter, the larger the volume that can be processed, but at reduced intensity
Specifications
Product Type
Digital Disperser
Amplitude
75 μm
Diameter (Metric)
6 mm
Packaging Quantity
1 Ea.