1x1 image pixel for data collection
twin.tec™ PCR Plates
twin.tec™ PCR Plates
Actual product may vary
Fisherbrand
twin.tec™ PCR Plates
Use these plates for automation and high throughput processes as well as quantitative real-time PCR. Eppendorf™ twin.tec™ PCR Plates combine extremely thin walled polypropylene wells for optimal heat transfer and extremely robust polycarbonate frames for rigidity and torque-resistance.
Have Questions?
Catalog NumberColorSkirt Style
FKS10120092ClearSkirted
Catalog number FKS10120092
Price (SGD)
878.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Color:
Clear
Skirt Style:
Skirted
Price (SGD)
878.00
PK
Add to cart
Use these plates for automation and high throughput processes as well as quantitative real-time PCR. Eppendorf™ twin.tec™ PCR Plates combine extremely thin walled polypropylene wells for optimal heat transfer and extremely robust polycarbonate frames for rigidity and torque-resistance.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Exceptionally solid and torque-resistant polycarbonate frame
  • Improved well-to-well tolerance
  • Cut-away corner and alphanumeric grid-referencing
  • Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors
Specifications
FormatStandard
Array8 x 12
AutoclavableAutoclavable
Packaging25 Pk.
Product LineEppendorf
ColorClear
DisposableSingle-use
Product TypePCR Microplate
For Use With (Equipment)Real time PCR
Skirt StyleSkirted
MaterialPolycarbonate
No. of Wells96
SterilitySterile
Total Volume (Metric) Well150 μL
Well ShapeConical Bottom (V-well)
Dnase FreeYes
For Use With (Application)Ideal for quantitative real time PCR and fit most thermal cyclers
Quantity25/Pk.
RNase FreeYes
Unit SizePK
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link