TC-Treated Culture Dishes
TC-Treated Culture Dishes
Actual product may vary
Fisherbrand
TC-Treated Culture Dishes
Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaShapeGriddedNo. per Pack
FKS10117320148 cm2RoundNot Gridded5/Pk.
FKS1009538155 cm2RoundNot Gridded10/Pk.
FIS08-772-209 cm2RoundNot Gridded20/Pk.
FIS08-772-2121 cm2RoundNot Gridded20/Pk.
FIS08-772-24148 cm2RoundNot Gridded5/Pk.
Catalog number FKS10117320
Price (SGD)
1,020.00
PK
Add to cart
Culture Area:
148 cm2
Shape:
Round
Gridded:
Not Gridded
No. per Pack:
5/Pk.
Price (SGD)
1,020.00
PK
Add to cart
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment
  • Ultra-Low Attachment surface dishes feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation
  • 10 dishes/bag are available for 100 mm dishes (Cat. No. 430293)
  • 245 mm square dishes offer 500 cm2 growth surface
  • Stacking beads aid in handling
  • Vents provide consistent gas exchange
  • Manufactured from optically clear virgin polystyrene
  • Sterile
  • Nonpyrogenic
Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.
Cat. no. 431110, Square dishes with interior bottom dimensions of 224 x 224 mm.
Specifications
Closure ColorClear
Closure IncludedYes
Closure MaterialPolystyrene
Bottom ShapeFlat
Capacity (Metric)45 mL (Well)
ColorClear
Culture Area148 cm2
DisposableSingle-use
GriddedNot Gridded
MaterialPolystyrene
ShapeRound
SterilitySterile
Surface TreatmentCell Culture
TypeCell Culture Petri Dish
Volume (Metric) Working30 to 45 mL
IncludesLid
No. per Case60/Cs.
No. per Pack5/Pk.
Size150 mm dia.
Height (Metric)25 mm
Diameter (Metric)150 mm
Unit SizePK