Zinc Alloy Bossheads
Actual product may vary
Fisherbrand
Zinc Alloy Bossheads
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Zinc Alloy Bossheads come with plated clamping screws with plastic heads.
Have Questions?
Catalog NumberSize
FKS1011718416 mm dia.
Catalog number FKS10117184
Price (SGD)
129.00
Pack
Size:
16 mm dia.
Price (SGD)
129.00
Pack
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Zinc Alloy Bossheads come with plated clamping screws with plastic heads.
  • For rods up to 16mm in diameter
  • Supplied in packs of 5
Specifications
Quantity5/Pk.
TypeBosshead Clamp
MaterialZinc Alloy
IncludesPlastic headed clamping screws
For Use With (Equipment)Rod up to 16 mm dia.
Size16 mm dia.
Unit SizePack