Matrix™ 500μL ScrewTop Tubes
Matrix™ 500μL ScrewTop Tubes
Actual product may vary
Matrix™ 500μL ScrewTop Tubes
Matrix™ 500μL ScrewTop Tubes
Matrix™ 500μL ScrewTop Tubes
Matrix™ 500μL ScrewTop Tubes
Matrix™ 500μL ScrewTop Tubes
Store samples securely at low temperatures, including vapor phase Liquid Nitrogen with these ScrewTop tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure ColorPackagingProduct Type
FKS10111683Colorless5 Latch Racks of 96 TubesScrew Top Tube
3743
also known as WM374X3A
ColorlessBulkScrew Top Tube
3744Colorless5 Latch Racks of 96 TubesScrew Top Tube
3744BLUBlue5 Latch Racks of 96 TubesScrew Top Tube
Catalog number FKS10111683
Price (SGD)
2,310.00
PK
Add to cart
Closure Color:
Colorless
Packaging:
5 Latch Racks of 96 Tubes
Product Type:
Screw Top Tube
Price (SGD)
2,310.00
PK
Add to cart
Store samples securely at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 500μL ScrewTop tubes. These tubes are molded from medical grade resins and feature a permanent, high contrast 2D barcode. Used in combination with Thermo Scientific™ VisionMate™ 2D barcode readers, manual and automated capping equipment, and labeling options, the 2D ScrewTop Tubes meet a range of manual and automated long term storage needs.
Dependable Sample Protection
  • Tubes are constructed of virgin class VI medical grade polypropylene
  • ScrewTop caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • ScrewTop Tubes are compatible with storage down to -176°C
Secure Tracking
  • Permanent, unique 2D barcode is laser-etched on the bottom of each tube
  • 2D barcode allows you to scan and decode tubes at once without removing them from the rack, and allows data to be associated with each tube
  • Compatible with our range of VisionMate readers
Superior Storage Format
  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping / decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at a time
Stringent Quality Control
  • Every 2D barcode is scanned for readability
  • Every 2D barcode is checked against the database of all previous Thermo Scientific 2D barcodes to ensure there are zero duplicates
  • Each tube is leak tested to ensure tube integrity and sample security
  • All tubes are supplied free from DNA, RNase, DNase, PCR Inhibitors, endotoxins and cytotoxins
Specifications
Product TypeScrew Top Tube
Closure ColorColorless
AutoclavableNot Autoclavable
Capacity (Metric)500 μL
Bottom ShapeConical
MaterialPolypropylene (Medical Grade)
Packaging5 Latch Racks of 96 Tubes
Well ShapeV-bottom
BarcodeNon-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction
Product LineMatrix™
SterilitySterile
Packaging Quantity480 Tubes
Closure IncludedYes
Closure MaterialVirgin Class VI Medical Grade Polypropylene with Virgin Class VI Medical Grade Silicone O-ring
Capacity (English)16907 fl oz.
Dnase FreeYes
Closure TypeColorless Cap
Fit for PurposeSterile and PCR Ready
Unit SizePK