Check Order Status
Volac Borosilicate Glass Class A Burette
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Volac Borosilicate Glass Class A Burette
Have Questions?
Contact Us
Catalog Number
Capacity (Metric)
FKS10108051
100 mL
Catalog number
FKS10108051
Price (SGD)
407.00
EA
Add to cart
Capacity (Metric):
100 mL
Price (SGD)
407.00
EA
Add to cart
Specifications
Color
Blue
Capacity (Metric)
100 mL
Length (Metric)
870 mm
Material
Borosilicate Glass
Graduations
0.2 mL
Includes
Interchangeable PTFE Stopcock
Scale
White
Stopcock
Straight
Stopcock Material
Glass
Tolerance
±0.1 mL
Type
Volumetric Buret
Class
Class AS
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link