Fluoropore™ Membrane Filters, 0.5 μm
Fluoropore™ Membrane Filters, 0.5 μm
Actual product may vary
Fisherbrand
Fluoropore™ Membrane Filters, 0.5 μm
Hydrophobic membrane with laminated backing for easier handling
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS10106190142 mm
FISFHLP14250142 mm
Catalog number FKS10106190
Price (SGD)
6,632.00
PK
Add to cart
Diameter (Metric):
142 mm
Price (SGD)
6,632.00
PK
Add to cart
Our Fluoropore™ membrane is a hydrophobic, polytetrafluoroethylene (PTFE) polymer membrane bonded to a high density polyethylene support to improve the handling characteristics of the filter for normal applications. Fluoropore™ membranes provide broad chemical compatibility, high flow rates and consistency. (Please note that catalogue numbers containing in.FHUP in. are unlaminated, or produced without a backing.)Applications: Fluoropore™ Membrane: Clarifying Acids, Bases and Solvents; Air Monitoring; Filtering and Venting Gases; UV Spectroscopy
Specifications
AutoclavableNot Autoclavable
Bubble Point7 psi
ColorWhite
Filter TypeScreen Filter
MaterialPTFE
Pore Size0.45 μm
SterilityNon-sterile
Flow Rate60 mL/min x cm2
For Use With (Application)Clarification of Acids, Bases and Solvents; Clarification of Alcohol Solutions; UV Spectroscopy.
Gas Flow Rate8.0 L/min/cm2
HydrophobicityHydrophobic
No. per Case50Pack
Quantity50/Pk.
TypeFilter Membranes
WettabilityHydrophobic
Diameter (Metric)142 mm
Unit SizePK