DuraCal™ 9.21 Value pH Buffer, Hamilton™
DuraCal™ 9.21 Value pH Buffer, Hamilton™
Fisherbrand
DuraCal™ 9.21 Value pH Buffer, Hamilton™
Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 9.21 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.
Have Questions?
Catalog NumberQuantity
FKS10092452500 mL
Catalog number FKS10092452
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
pH9.21
Quantity500 mL
Unit SizeEA
  • 9.21 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards