Check Order Status
Air Condensers
Actual product may vary
Fisherbrand
Air Condensers
Simple condensers for condensation of materials with boiling points above 150°C.
Have Questions?
Catalog Number
Type
FKS10089109
Air Condenser
Catalog number
FKS10089109
Price (SGD)
104.00
EA
Add to cart
Type:
Air Condenser
Price (SGD)
104.00
EA
Add to cart
Simple but effective
Plain column
Ground socket
Cone joints
Specifications
Description
Socket size 19/26
Length (Metric)
150 mm
Quantity
Each
Standard Taper Joint No
19/26
Material
Borosilicate Glass
Type
Air Condenser