Check Order Status
Natural Rubber Safety Pipette Fillers
Actual product may vary
Fisherbrand
Natural Rubber Safety Pipette Fillers
Made of natural rubber
Have Questions?
Catalog Number
Type
Model
FKS10079780
Safety Pipette Filler
Universal
Catalog number
FKS10079780
Price (SGD)
41.00
EA
Add to cart
Type:
Safety Pipette Filler
Model:
Universal
Price (SGD)
41.00
EA
Add to cart
Universal use for volumetric and graduated pipettes
Specifications
Color
Red
Material
Natural Rubber
Model
Universal
Type
Safety Pipette Filler
For Use With (Equipment)
volumetric and graduated pipettes