Specifications
ColorTransparent
Capacity (Metric)60 mL
MaterialPolypropylene
SterilityNon-sterile
TypeReagent Reservoir
For Use With (Application)For use with multichannel pipets. Includes lid.
Height (English)2.8 in.
Height (Metric)7.11 cm
IncludesLid
Length (English)5.5 in.
Length (Metric)13.97 cm
PackagingPacks of 10
Quantity10/Pk.
Width (English)4.6 in.
Width (Metric)11.68 cm
AutoclavableAutoclavable
Channels1
DisposableLimited-use
Bottom ShapeV-bottom
Unit SizePK