Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Actual product may vary
Fisher Catalogue
Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: Without Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsNo. per PackNo. per Case
FKS10058820961 Microplate50 Microplates
FIS12-565-64961 Microplate50 Microplates
Catalog number FKS10058820
View Price:Sign in to see your account pricing. Need an account? Register with us today.
No. of Wells:
96
No. per Pack:
1 Microplate
No. per Case:
50 Microplates
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWithout Lid
Description96F well plate, Nunclon Delta, clear, without lid, Sterile (Pack of 1, Case of 50)
Array8 x 12
Packaging1/Pk., 50/Cs.
Product LineNunc
ColorClear
Product TypeMicroplate
Skirt StyleHigh Flange
MaterialPolystyrene
Volume (Metric) Well400 μL
No. of Wells96
SterilitySterile
Surface TreatmentNunclon Delta
Total Volume (Metric) Well400 μL
Well ShapeFlat Bottom (F-well)
No. per Case50 Microplates
No. per Pack1 Microplate
Quantity50/Cs.
Unit SizePK