Finntip™ Universal Pipette Tips
Finntip™ Universal Pipette Tips
Actual product may vary
Fisher Catalogue
Finntip™ Universal Pipette Tips
Ensure accurate, precise pipetting with these universal pipette tips, designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Have Questions?
Catalog NumberDescriptionVolume (Metric)FormatSterility
FKS10053860Finntip 250 Universal, Non-Filtered, Non-sterile, Box250 μLBoxNon-sterile
Catalog number FKS10053860
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Finntip 250 Universal, Non-Filtered, Non-sterile, Box
Volume (Metric):
250 μL
Format:
Box
Sterility:
Non-sterile
Ensure accurate, precise pipetting with Thermo Scientific™ Finntip™ Universal Pipette Tips, manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips to prevent liquid retention.
  • Smooth, uniform surface prevents liquid retention
  • Volume range from 0.2μL to 10mL
  • Available sterile or nonsterile
  • Packaging options include racks, bags, boxes, bulk, and refill packs
  • Color-coded racks to match the compatible Finnpipette
  • Color-coding only applies to rack and refill packaging
Note:
Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified)
Specifications
DescriptionFinntip 250 Universal, Non-Filtered, Non-sterile, Box
Volume (Metric)250 μL
FilteredNon-filtered
SterilityNon-sterile
ColorYellow
ModelFinntip 250 Universal
Length (English)2 in.
Length (Metric)5.2cm
Certifications/ComplianceISO 9001, ISO 14001, ISO 13485
FormatBox
Tip StyleUniversal Fit
Packaging500 tips
For Use With (Equipment)Thermo Scientific™ Finnpipette™ 250
Product LineFinnTip
Unit SizePK