Orion™ Temperature Probes
Orion™ Temperature Probes
Actual product may vary
Fisherbrand
Orion™ Temperature Probes
Automatically correct pH measurements for variation in electrode slope due to temperature change.
Have Questions?
Catalog NumberConnector TypesType
FKS10050113Mini-DINATC Probe
Catalog number FKS10050113
Price (SGD)
1,674.00
EA
Add to cart
Connector Types:
Mini-DIN
Type:
ATC Probe
Price (SGD)
1,674.00
EA
Add to cart
Automatically correct pH measurements for variation in electrode slope due to temperature change using Thermo Scientific™ Orion™ Stainless-Steel Automatic Temperature Compensation (ATC) Probes.
Temperature Range: 0° to 100°C 
Relative Accuracy: ±1°C
pH Range: 0 to 14
Storage: Store in air
Includes: 1m cable
Note:  The connector must be compatible with the meter you are using.
Warranty: 12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier.
Specifications
Accuracy±2°C
For Use With (Equipment)Orion Star, Star A, Versa Star
MaterialStainless Steel
Temperature (Metric) Operating0°C to 100°C
TypeATC Probe
DescriptionATC probe with stainless-steel body
Diameter (Metric) Body6 mm
Length (Metric)120 mm
Temperature (English) Operating32°F to 212°F
Connector TypesMini-DIN
Storage RequirementsStore in air
Unit SizeEA