PMMA Semi-Micro Cuvettes
PMMA Semi-Micro Cuvettes
Actual product may vary
Fisherbrand
PMMA Semi-Micro Cuvettes
These macro or semi-micro cuvettes that are available in PS and PMMA are ideal for kinetic measurements.
Have Questions?
Catalog NumberCapacity (Metric)
FKS100388101.5 to 3 mL
Catalog number FKS10038810
Price (SGD)
80.00
PK
Add to cart
Capacity (Metric):
1.5 to 3 mL
Price (SGD)
80.00
PK
Add to cart
  • Macro or semi-micro cuvettes available in PS and PMMA
  • Ideal for kinetic measements
  • Windows are recessed to protect from scratches, arrow mark indicates optical path orientation
  • Plastibrand cuvettes are delivered grouped by mold cavity number to ensure lowest variation of extinction coefficient
  • Useable wavelength > 300 nm
  • Standard deviation at 320 nm: ±0.004 extinction units
Specifications
TypeSemi-micro Cuvet
Capacity (Metric)1.5 to 3 mL
MaterialPMMA
Quantity100Pack
Unit SizePK