Search Fisher Scientific
| Catalog Number | Diameter (Metric) | Color | No. per Case |
|---|---|---|---|
| FKS10034410 | 47 mm | White | 100 Pk. |
| FKS10529202 | 25 mm | White | 100 Pk. |
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features and Benefits: