Modified 7000 Microliter™ Syringes
Modified 7000 Microliter™ Syringes
Actual product may vary
Fisherbrand
Modified 7000 Microliter™ Syringes
This series of syringes is ideal for dispensing volumes from 0.05 μL up to 5 μL. The 7000 series is the only zero dead volume syringe. The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle GaugeNeedle Point NumberNeedle Point Style
FKS100284201 μL25 G2Sharp, Bevel, Curved, Non-coring
FIS14-813-1000.5 μL25 G3Blunt
FIS14-824-201 μL25 G3Blunt
FIS14-824-27C1 μL25 G2Sharp, Bevel, Curved, Non-coring
Catalog number FKS10028420
Price (SGD)
450.00
EA
Add to cart
Volume (Metric):
1 μL
Needle Gauge:
25 G
Needle Point Number:
2
Needle Point Style:
Sharp, Bevel, Curved, Non-coring
Price (SGD)
450.00
EA
Add to cart
  • ideal for dispensing volumes from 0.05 µL up to 5 µL
  • The 7000 series is the only zero dead volume syringe
  • The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
AutoclavableAutoclavable
Needle Gauge25 G
Diameter (English) Inner0.005 in.
Diameter (English) Inner Needle0.005 in.
Diameter (Metric) Inner Needle0.14 mm
Diameter (English) Outer Needle0.02 in.
Diameter (English) Outer0.3 in.
Diameter (Metric) Inner0.14 mm
Diameter (Metric) Outer7.82 mm
Diameter (Metric) Outer Needle0.515 mm
Length (English) Needle2.75 in.
Product TypeGeneral Purpose Syringe
Max. Pressure6000 psi
Max. Temperature (English)239°F
Max. Temperature (Metric)115°C
Min. Temperature (English)50°F
Needle Point Number2
Min. Temperature (Metric)10°C
QuantityEa.
Volume (Metric)1 μL
Needle Point StyleSharp, Bevel, Curved, Non-coring
Connection TypeKnurled Hub, Removable
Unit SizeEA