1x1 image pixel for data collection
Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts
Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts
Actual product may vary
Fisherbrand
Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts
Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeNo. of WellsDiameter (Metric) OuterCulture Area
FKS100254713 μm246.5 mm0.33 cm2
FIS07-200-1650.4 μm624.5 mm4.7 cm2
FIS07-200-1573 μm1212 mm1.12 cm2
FIS07-200-1495 μm246.5 mm0.33 cm2
FIS07-200-1470.4 μm246.5 mm0.33 cm2
Catalog number FKS10025471
Price (SGD)
2,515.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Pore Size:
3 μm
No. of Wells:
24
Diameter (Metric) Outer:
6.5 mm
Culture Area:
0.33 cm2
Price (SGD)
2,515.00
PK
Add to cart
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
You can use cell culture inserts to:
  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
  • Pore sizes ranging from 0.4 μm to 8 μm diameters
  • Treated for optimal cell attachment
  • Supplied in multiple well plates
  • Membrane must be stained for cell visibility
  • Sterile
Specifications
Description12 Inserts
Culture Area0.33 cm2
MaterialPolycarbonate
Volume (Metric) Well0.6 mL
Pore Size3 μm
TypePre-loaded Microporous Insert in Tissue Culture Plate
For Use With (Application)Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies
Diameter (Metric) Outer6.5 mm
No. per Pack4 plates; 12 inserts/plate
Surface CoatingTC-Treated
ColorClear
Surface TreatmentTC-Treated
SterilitySterile
No. of Wells24
Unit SizePK
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection