Digital Hand-Held Pocket Refractometer: PAL-RI
Digital Hand-Held Pocket Refractometer: PAL-RI
Actual product may vary
Digital Hand-Held Pocket Refractometer: PAL-RI
Digital Hand-Held Pocket Refractometer: PAL-RI
Fisherbrand
Digital Hand-Held Pocket Refractometer: PAL-RI
nD 1.3306-1.5284. The ATAGO™ Digital Hand-Held Pocket Refractometer: PAL-RI provides an accuracy of ±0.0003 (water at 20°C). When the start key is pressed after placing some droplets of sample on the prism, the refractive index (resolution 0.0001 ) will be displayed continuously like an electric newsboard.
Have Questions?
Catalog number FKS10014082
Price (SGD)
1,719.00
EA
Add to cart
Price (SGD)
1,719.00
EA
Add to cart
Measurement time: 3 seconds.
Specifications
TypeRefractometer
Refractive Index1.3306 to 1.5284
ModelPAL-RI
Sample Volume0.3 mL
Accuracy±0.0003 (water at 20°C)
Resolution0.0001
Scale TypeRefractive Index
Measurement Time3 sec.
IP RatingIP65 Dust-tight and Protected against water jets
Weight (Metric)100 g
Ambient Temperature (Metric)10°C to 40°C
Temperature CompensationNo
Power Supply Details2xAAA Batteries
Unit SizeEA