Cautery Pens, High Temperature
Cautery Pens, High Temperature
Actual product may vary
Fisher Catalog
Cautery Pens, High Temperature
Stoelting™ High Temperature Cautery Pens are popular for procedures including pinpoint hemostasis, controlling diffuse bleeders, vasectomies, and sculpting woven grafts.
Have Questions?
Catalog NumberModelTemperature (Metric) Range
FKS10001044Vasectomy micro tip871°C
Catalog number FKS10001044
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Model:
Vasectomy micro tip
Temperature (Metric) Range:
871°C
  • Sterile and disposable
  • Stop bleeding immediately with these handy, sterile, disposable cautery pens.
  • Platinum filaments reach up to 2200°F (low-temp pen up to 850°F) within milliseconds, to heat-seal cut blood vessels.
  • Tip allows positioning without obstructing the user's view.
  • Locking cap secures the on/off switch to prevent accidental operation, and protects the tip.
  • Discard the instrument when the built-in batteries are dead.
  • Supplied in packages of 10
  • Each pen is packaged in it's own sterile pouch
Specifications
For Use With (Application)Neuroscience
Temperature (Metric) Range871°C
ModelVasectomy micro tip
Unit SizePK