NERL™ Trutol™ Glucose Tolerance Test Beverages
NERL™ Trutol™ Glucose Tolerance Test Beverages
Actual product may vary
Thermo Scientific
NERL™ Trutol™ Glucose Tolerance Test Beverages
Screen for diabetes mellitus, gestational diabetes and hypoglycemia with these glucose tolerance test beverages.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFlavorDose
FISTGP401223PAOrange75g
FISTGP401074PALemon-Lime50g
FISTGP401526PFruit Punch75g
FISTGP401025PALemon-Lime75g
Catalog number FISTGP401223PA
Price (SGD)
-
Flavor:
Orange
Dose:
75g
Screen for diabetes mellitus, gestational diabetes and hypoglycemia with Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages. Beverages are noncarbonated, caffeine-free and available in 50, 75 and 100g concentrations.
  • Noncarbonated beverages provide easier consumption, a reduction of digestive discomfort and a more palatable taste
  • Graduated bottles assure proper dose every time
  • Kosher-certified
Ordering Alerts:
  • Plastic bottles packaged in cases of 24 bottles; four six-packs per case
Specifications
Certifications/ComplianceKosher
Detectable AnalytesNone
TypeReagent
Dose75g
FlavorOrange
Packaging Type24 Bottles/Cs.
Quantity10 oz.
Shelf Life24 Months
Unit SizeCase
Contents & Storage
15°C to 30°C