Check Order Status
Sodium Hydroxide Solution (0.25N (N/4)/Certified), Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide Solution (0.25N (N/4)/Certified), Fisher Chemical™
A volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
FISSS272-1
1 L
Poly Bottle
Catalog number
FISSS272-1
Price (SGD)
-
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Hydroxide Solution
pH
13
Cleanup Symbol
Caustic
Color
Colorless
Name Note
0.25N (N/4)
View more