Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane
Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane
Actual product may vary
MilliporeSigma
Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane
Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions are available in 0.1, 0.22, 0.45 and 5.0μm pore sizes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore Size
FISSLVVM33RS0.1 μm
FISSLGVM33RS0.22 μm
Catalog number FISSLVVM33RS
Price (SGD)
777.00
PK
Add to cart
Pore Size:
0.1 μm
Price (SGD)
777.00
PK
Add to cart
Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions are available in 0.1, 0.22, 0.45 and 5.0μm pore sizes
  • 100% integrity tested, individually packaged, and sterilized
  • Female Luer-Lok™ inlet and male Luer slip outlet
  • Manufactured in a controlled environment using an automated process
  • Yellow color-coded band clearly indicates which membrane is inside
  • ≤0.22μm filters are sterilizing grade
  • Larger pore sizes are for clarification and prefiltration
  • Include a Certificate of Quality
Durapore™ (PVDF) membrane
  • Lowest protein binding sterilization/clarification of biological solutions
  • Made of polyvinylidene fluoride (PVDF)
  • Gamma irradiation sterilized
Compliance
  • Meet USP Class VI Biological tests for plastics
  • All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Specifications
AutoclavableNot Autoclavable
Certifications/ComplianceMeets USP Class VI Biological tests for plastics. All 33 mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
ColorYellow
DisposableSingle-use
Material (Membrane)PVDF (Hydrophilic)
Pore Size0.1 μm
SterilitySterile
Filter CodeVV
For Use With (Application)Ultra-low protein binding; Sterilization of tissue culture media, protein solutions or aqueous solutions
Hold up Volume (After Air Purge)<100 μL
Housing MaterialPVDF
Inlet StyleFemale Luer Lock
Outlet StyleMale Luer Slip
Pressure Rating145 psi
Quantity50/Pk.
Temperature (English) Operating113°F
Temperature (Metric) Operating45°C
Temperature Range (English)113°F
Temperature Range (Metric)45°C
Tip MaterialModified Acrylic
TypeSyringe Filter, Medical
Volume (Metric)10 mL
Diameter (Metric)3.3 cm
Unit SizePK