1x1 image pixel for data collection
Millex™ Syringe Filter, Durapore™ (PVDF), 0.45 μm
Millex™ Syringe Filter, Durapore™ (PVDF), 0.45 μm
Actual product may vary
MilliporeSigma
Millex™ Syringe Filter, Durapore™ (PVDF), 0.45 μm
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Have Questions?
Catalog NumberOutlet StyleQuantity
FISSLHVX13NLMale Luer Slip100/Pk.
Catalog number FISSLHVX13NL
Price (SGD)
940.00
PK
Estimated availability date 17-Sep-2025
Add to cart
Outlet Style:
Male Luer Slip
Quantity:
100/Pk.
Price (SGD)
940.00
PK
Add to cart
Overmolded polypropylene housing and optimized housing design
Overview of Non-Sterile Millex™ Syringe Filters with Durapore™ (PVDF) Membrane
  • Low protein binding to minimize interaction with your sample and maximize recovery
  • Compatible with aqueous and mild organic solutions
  • Housing materials are either polypropylene or high density polyethylene (HDPE), offering low levels of extractables and broad chemical compatibility
Specifications
Color CodeYellow
DisposableSingle-use
Material (Membrane)PVDF (Hydrophilic)
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)Analytical sample prep
Housing MaterialPP
Inlet StyleMale Luer Slip
Outlet StyleMale Luer Slip
Product LineMillex™
Quantity100/Pk.
TypeSyringe Filter
Volume (Metric)<10 mL
Diameter (Metric)13 mm
Diameter (Metric) Filter13 mm
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link