HyClone™ Minimum Essential Medium
HyClone™ Minimum Essential Medium
Actual product may vary
Fisher Catalogue
HyClone™ Minimum Essential Medium
Support cell culture growth and biomanufacturing processes with HyClone™ Minimum Essential Medium (MEM) with Earle's Balanced Salts (MEM/EBSS), manufactured based on compliance with ISO 9001:2000 guidelines and current Good Manufacturing Practices (cGMP) for Medical Devices.
Have Questions?
Catalog NumberQuantityWith Additives
FISSH30024021000 mLGlutamine, Earle's Balanced Salt Solution (EBSS)
Catalog number FISSH3002402
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
With Additives:
Glutamine, Earle's Balanced Salt Solution (EBSS)
  • System tested with Cytiva HyClone sera to ensure product efficacy and homogeneity
  • All products are hydrated using purified process water and undergo sterile filtration
  • Manufactured using ISO 9001 certified processes
Specifications
FormLiquid
Product TypeMEM with EBSS, L-glutamine
Shelf Life12 Months
SterilitySterile-filtered
Sterilization Method0.1 μm Sterile-filtered
With AdditivesGlutamine, Earle's Balanced Salt Solution (EBSS)
Without AdditivesNone
Quantity1000 mL
Certifications/CompliancecGMP, ISO 9001:2000
DescriptionMEM with Earle's Balanced Salt Solution (EBSS), with 2.0mM L-Glutamine
Unit SizeEA
Contents & Storage
2° to 8°C, away from light