Syringes with Valves
Actual product may vary
Syringes with Valves
Syringes with Valves
SGE
Syringes with Valves
Prefitted with removable needle and valve or Luer Lock valve
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FISSG00967050 mL
FISSG-00947225 mL
FISSG009770100 mL
Catalog number FISSG009670
Price (SGD)
-
Volume (Metric):
50 mL
  • Low syringe carryover
  • Reduced nonspecific sample interaction
  • Increased analyte recovery
  • Ideal for sample storage and transportation or to pressurize sample prior to injection
  • Material: Borosilicate glass, PTFE plunger (Gas tight)
With Removable Needle
  • Needle Length: 50mm (2 in.)
  • Gauge: 23
  • Outside Diameter: 0.63mm
  • Inside Diameter: 0.2mm
  • Cone Tip
Specifications
AutoclavableAutoclavable
MaterialBorosilicate Glass, PTFE (Plunger)
For Use With (Application)For sample storage and transportation or to pressurize sample prior to injection
Product TypeSyringe
Volume (Metric)50 mL
Connection TypeLuer Lock, Removable
Unit SizeEach