VSV-6 Vespel Valve Seat
Actual product may vary
Trajan
VSV-6 Vespel Valve Seat
Precision metering and on-off control of gas flows. Designed specifically for chromatography applications and may be used at elevated temperatures from high vacuum to medium pressure.
Have Questions?
Catalog NumberFor Use With (Equipment)
FISSG-123681Replacement for VSV-6 Micro Valves
Catalog number FISSG-123681
Price (SGD)
-
For Use With (Equipment):
Replacement for VSV-6 Micro Valves
  • Ideal for chromatography applications
  • May be used at elevated temperatures from high vacuum to medium pressure
  • Vacuum rated to a leak rate of 1E-05 cc/sec of helium
Specifications
TypeValve Seat, Vespel
StyleVSV-6 SGE
For Use With (Equipment)Replacement for VSV-6 Micro Valves
Unit SizePack