Check Order Status
Buffer Solution, pH 2.00 (Certified), Fisher Chemical™
Element Search
Structure Search
Buffer Solution, pH 2.00 (Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FISSB96-500
500 mL
Poly Bottle
Catalog number
FISSB96-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7, 7647-01-0, 50-00-0, 67-56-1, 7732-18-5
Specifications
Chemical Name or Material
Buffer Solution, pH 2.00
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
Melting Point
0°C
Boiling Point
100°C
View more