Check Order Status
Buffer Solution, pH 6.00 (Certified), Fisher Chemical™
Element Search
Structure Search
Buffer Solution, pH 6.00 (Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FISSB104-500
500 mL
Poly Bottle
Catalog number
FISSB104-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7778-77-0, 7732-18-5, 1310-73-2
Specifications
Chemical Name or Material
Buffer Solution, pH 6.00
CAS
7778-77-0
ChemAlert Storage Symbol
Gray
Melting Point
0°C
Boiling Point
100°C
View more