Litmus Paper
Actual product may vary
United Scientific Supplies
Litmus Paper
Available as Litmus paper and Litmus Granular in bottles.
Have Questions?
Catalog NumberTypeQuantity
FISS99401Neutral Litmus Paper100 Strips/Vial
Catalog number FISS99401
Price (SGD)
-
Type:
Neutral Litmus Paper
Quantity:
100 Strips/Vial
  • 100 strips each in matching color plastic vials
  • Individual strips are approximately 2 in. long and 0.25 in. wide
Specifications
pH Range0 to 14
FormatPaper Strip
For Use With (Application)Measuring pH or acidity in liquids
Quantity100 Strips/Vial
TypeNeutral Litmus Paper
Dimensions (L x W)2 x 0.25 in. (5.08 x 0.635 cm)
Length (English)2 in.
Length (Metric)5.08 cm
Width (English)0.25 in.
Width (Metric)0.635 cm
Unit SizeEach