Fuel Cell Car Science Kit
Fuel Cell Car Science Kit
Actual product may vary
Fuel Cell Car Science Kit
Combines cutting-edge science, education, and fun for all
Have Questions?
Catalog NumberGrade
FISS986066+
Catalog number FISS98606
Price (SGD)
-
Grade:
6+
  • Uses a reversible PEM fuel cell that combines electrolysis and power conversion into one device
  • Oxygen and hydrogen gases are formed in two transparent water containers
  • Steers independently of the user once in operation
  • Runs past obstacles; if the car hits a barrier, it automatically reverses 90°
  • Grades: 6+
Renewable Energy Education CD
  • Renewable Energy Science Education Manual: Introduces renewable energy in seven chapters—Global Warming and Climate Change, Solar Technologies, Wind Technologies, Electrolyzers, Fuel Cells, Hydrogen Storage, and Basic Electronics
  • Experiment Manual: More than 20 experiments for middle to high school level sciences in several sections—Solar Energy, Wind Energy, Energy from Hydrogen (Fuel Cell), and “Ultra Cool« experiments
  • Flash Animations: Describe and show how a fuel cell works, different types of fuel cells, electrolysis, and various applications for fuel cells
  • Assembly Guides
  • Technical Support with videos
Specifications
Product TypeFuel Cell Car Science Kit
Grade6+
For Use With (Application)Demonstrates electrolysis of water into oxygen and hydrogen
For Use With (Equipment)Electrolysis and power conversion into one single device
IncludesCar, solar panel, water-filling syringe, all necessary components, instruction manual and a renewable energy education CD
Unit SizeEA